missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NU 2058, TCI America™

$218.48 - $763.91

Chemical Identifiers

CAS 161058-83-9
Molecular Formula C12H17N5O
Molecular Weight (g/mol) 247.302
MDL Number MFCD05664734
InChI Key MWGXGTJJAOZBNW-UHFFFAOYSA-N
Synonym 6-cyclohexylmethoxy-9h-purin-2-amine, 6-o-cyclohexylmethyl guanine, 2-amino-6-cyclohexylmethoxypurine, 2-amino-6-cyclohexylmethyl oxy purine, 6-cyclohexylmethoxy-7h-purin-2-amine, 9h-purin-2-amine, 6-cyclohexylmethoxy, o-cyclohexylmethylguanine, o6-cyclohexylmethylguanine, d0r1ft, d0sj6y
PubChem CID 4564
IUPAC Name 6-(cyclohexylmethoxy)-7H-purin-2-amine
SMILES C1CCC(CC1)COC2=NC(=NC3=C2NC=N3)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N0958100MG View Documents TCI America
N0958100MG
100mg Each for $763.91
 
N095820MG View Documents TCI America
N095820MG
20mg Each for $218.48
 
Specifications

Chemical Identifiers

161058-83-9
247.302
MWGXGTJJAOZBNW-UHFFFAOYSA-N
4564
C1CCC(CC1)COC2=NC(=NC3=C2NC=N3)N
C12H17N5O
MFCD05664734
6-cyclohexylmethoxy-9h-purin-2-amine, 6-o-cyclohexylmethyl guanine, 2-amino-6-cyclohexylmethoxypurine, 2-amino-6-cyclohexylmethyl oxy purine, 6-cyclohexylmethoxy-7h-purin-2-amine, 9h-purin-2-amine, 6-cyclohexylmethoxy, o-cyclohexylmethylguanine, o6-cyclohexylmethylguanine, d0r1ft, d0sj6y
6-(cyclohexylmethoxy)-7H-purin-2-amine

Specifications

White-Yellow
100mg
C12H17N5O
6-cyclohexylmethoxy-9h-purin-2-amine, 6-o-cyclohexylmethyl guanine, 2-amino-6-cyclohexylmethoxypurine, 2-amino-6-cyclohexylmethyl oxy purine, 6-cyclohexylmethoxy-7h-purin-2-amine, 9h-purin-2-amine, 6-cyclohexylmethoxy, o-cyclohexylmethylguanine, o6-cyclohexylmethylguanine, d0r1ft, d0sj6y
C1CCC(CC1)COC2=NC(=NC3=C2NC=N3)N
247.302
247.30
≥98.0% (HPLC)
199°C
161058-83-9
MFCD05664734
MWGXGTJJAOZBNW-UHFFFAOYSA-N
6-(cyclohexylmethoxy)-7H-purin-2-amine
4564
Crystal-Powder at 20°C
NU 2058
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator