Please login to your online account to display your discounted pricing

NU 1025, Tocris

Potent PARP inhibitor

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material NU 1025
Assay Percent Range >99%
Molecular Formula C9H8N2O2
Formula Weight 176.17
Synonym 8-Hydroxy-2-methyl-4(3H)-quinazolinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

140110

 
r & d systems
140110
10mg Each for $188.60

140150

 
r & d systems
140150
50mg Each for $794.96
Description & Specifications

Specifications

Chemical Name or Material NU 1025
Quantity 10mg
CAS 90417-38-2
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C9H8N2O2
Formula Weight 176.17
Synonym 8-Hydroxy-2-methyl-4(3H)-quinazolinone
Safety and Handling

Store at Room Temperature