Please login to your online account to display your discounted pricing

Tocris™ nTZDpa

Potent, selective PPARgamma partial agonist

$212.74 - $886.68

Specifications

Product Type nTZDpa
Inhibitors nTZDpa
Molecular Weight 428.33
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

215010R

 
tocris™
215010
10mg Each for $212.74

215050

 
tocris™
215050
50mg Each for $886.68
Description & Specifications

Specifications

Product Type nTZDpa
Inhibitors nTZDpa
Molecular Weight 428.33

Potent, selective non-thiazolidinedione PPARγ partial agonist (EC50 = 57nM); produces ∽25% maximum efficacy. Antagonizes full agonist activity by ∽60% (IC50 ∽ 285nM). Displays no activity at PPARα or PPARδ receptors. Produces altered receptor conformation, and regulates adipocyte development and gene expression, in a differential manner to full PPARγ agonists. Modulates metabolism and insulin sensitivity without causing cardiac hypertrophy in mice in vivo.

  • Chemical Name: 5-Chloro-1-[(4-chlorophenyl)methyl]-3-(phenylthio)-1H-indole-2-carboxylic acid
  • Formula: C22H15Cl2NO2S
  • CAS Number: 118414-59-8
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >98