missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NTRK3 (TRKC) Recombinant Human Protein DFS Item

NTRK3 (TRKC) Recombinant Human Protein

$576.00 - $576.00

Specifications

Accession Number NP_002521
Assays NTRK3 (TRKC) was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Common Name NTRK3 (TRKC) Recombinant Human Protein
Expression System Baculovirus
Gene Alias NTRK3 (TRKC)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV3617 View Documents Life Technologies
LSPV3617
10μg Each for $576.00
 
Description

Description

NTRK3 (TRKC) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system.
Specifications

Specifications

NP_002521
NTRK3 (TRKC) Recombinant Human Protein
NTRK3 (TRKC)
41.2kDa
1 Each
Amino acids 510-825
For research use only. Not for use in diagnostic procedures
Kinase assay
His Tag
NTRK3 (TRKC) was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Baculovirus
Neurotrophic receptor (NTRK)
NTRK3 (TRKC) Recombinant Human Protein
NTRK3 (TRKC) Recombinant Human Protein
80% by SDS-PAGE
–80°C
Human

For Research Use Only. Not for use in diagnostic procedures.