Please login to your online account to display your discounted pricing

NSC 74859, Tocris™

Selective STAT3 inhibitor

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material NSC 74859
Assay Percent Range >98%
Molecular Formula C16H15NO7S
Formula Weight 365.36
Synonym 2-Hydroxy-4-[[2-[[(4-methylphenyl)sulfonyl]oxy]acetyl]amino]benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

465510

 
r & d systems
RDS465510
10mg Each for $188.60

465550

 
r & d systems
RDS465550
50mg Each for $794.96
Description & Specifications

Specifications

Formula Weight 365.36
Quantity 10mg
Molecular Formula C16H15NO7S
Assay Percent Range >98%
Synonym 2-Hydroxy-4-[[2-[[(4-methylphenyl)sulfonyl]oxy]acetyl]amino]benzoic acid
Chemical Name or Material NSC 74859
CAS 501919-59-1
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C