missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NSC 693868, Tocris

Cdk inhibitor; Also inhibits GSK-3

$244.81 - $997.23

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C9H7N5
Formula Weight 185.19
Synonym 1H-Pyrazolo[3, 4-b]quinoxalin-3-amine
Chemical Name or Material NSC 693868
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
193710 R & D Systems
193710
10mg Each for $244.81
193750 R & D Systems
193750
50mg Each for $997.23
Specifications Description & Specifications

Chemical Identifiers

>99%
185.19
NSC 693868
C9H7N5
1H-Pyrazolo[3, 4-b]quinoxalin-3-amine

Specifications

10mg
>99%
185.19
NSC 693868
40254-90-8
C9H7N5
1H-Pyrazolo[3, 4-b]quinoxalin-3-amine

Safety and Handling

Recommended Storage : Store at +4°C