Please login to your online account to display your discounted pricing

NSC 687852, Tocris™

Inhibitor UCHL5 and USP14

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material NSC 687852
Assay Percent Range >97%
Molecular Formula C22H17N3O6
Formula Weight 419.39
Synonym 3, 5-Bis[(4-nitrophenyl)methylene]-1-(1-oxo-2-propene-1-yl)-4-piperidone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

456610

 
r & d systems
RDS456610
10mg Each for $181.19

456650

 
r & d systems
RDS456650
50mg Each for $762.58
Description & Specifications

Specifications

Formula Weight 419.39
Quantity 10mg
Chemical Name or Material NSC 687852
Assay Percent Range >97%
Molecular Formula C22H17N3O6
CAS 1009817-63-3
Synonym 3, 5-Bis[(4-nitrophenyl)methylene]-1-(1-oxo-2-propene-1-yl)-4-piperidone
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C