missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NSC 66811, Tocris

MDM2 antagonist; Disrupts MDM2-p53 interaction

$218.74 - $915.10

Chemical Identifiers

Assay Percent Range >97%
Molecular Formula C23H20N2O
Formula Weight 340.42
Synonym 2-Methyl-7-[Phenyl(phenylamino)methyl]-8-quinolinol
Chemical Name or Material NSC 66811
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
293610 R & D Systems
RDS293610
10mg Each for $218.74
293650 R & D Systems
RDS293650
50mg Each for $915.10
Specifications Description & Specifications

Chemical Identifiers

>97%
340.42
NSC 66811
C23H20N2O
2-Methyl-7-[Phenyl(phenylamino)methyl]-8-quinolinol

Specifications

10mg
>97%
340.42
NSC 66811
6964-62-1
C23H20N2O
2-Methyl-7-[Phenyl(phenylamino)methyl]-8-quinolinol

Safety and Handling

Recommended Storage : Store at +4°C