Please login to your online account to display your discounted pricing

NSC 632839 hydrochloride, Tocris

Inhibitor of ubiquitin isopeptidase activity

$163.07 - $678.85

Chemical Identifiers

Chemical Name or Material NSC 632839 hydrochloride
Assay Percent Range >99%
Molecular Formula C21H21NO·HCl
Formula Weight 339.86
Synonym 3, 5-Bis[(4-methylphenyl)methylene]-4-piperidone hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

264710

 
r & d systems
RDS264710
10mg Each for $163.07

264750

 
r & d systems
RDS264750
50mg Each for $678.85
Description & Specifications

Specifications

Formula Weight 339.86
Quantity 10mg
Chemical Name or Material NSC 632839 hydrochloride
CAS 157654-67-6
Assay Percent Range >99%
Synonym 3, 5-Bis[(4-methylphenyl)methylene]-4-piperidone hydrochloride
Molecular Formula C21H21NO·HCl
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C