Please login to your online account to display your discounted pricing

Tocris™ NSC 625987

Cyclin-dependent kinase 4 (cdk4) inhibitor

$214.12 - $895.76

Specifications

Product Type NSC 625987
Inhibitors NSC 625987
Molecular Weight 271.33
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

215210R

 
tocris™
215210
10mg Each for $214.12

215250

 
tocris™
215250
50mg Each for $895.76
Description & Specifications

Specifications

Product Type NSC 625987
Inhibitors NSC 625987
Molecular Weight 271.33
Purity >98

Cyclin-dependent kinase (cdk) 4 inhibitor (IC50 = 0.2μM at cdk4/cyclin D1). Displays > 500-fold selectivity over cdk2 (IC50 > 100μM for cdc2/cyclin A, cdk2/cyclin A and cdk2/cyclin E).

  • Chemical Name: 1,4-Dimethoxy-9(10H)-acridinethione
  • Formula: C15H13NO2S
  • CAS Number: 141992-47-4
  • Solubility: Soluble to 10mM in ethanol and to 100mM in DMSO
  • Purity: >98