Please login to your online account to display your discounted pricing

NSC 405020, Tocris™

Inhibitor of MT1-MMP; antitumor

$139.08 - $585.69

Chemical Identifiers

Chemical Name or Material NSC 405020
Assay Percent Range >99%
Molecular Formula C12H15Cl2NO
Formula Weight 260.16
Synonym 3, 4-Dichloro-N-(1-methylbutyl)benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

490210

 
r & d systems
RDS490210
10mg Each for $139.08

490250

 
r & d systems
RDS490250
50mg Each for $585.69
Description & Specifications

Specifications

Formula Weight 260.16
Quantity 10mg
Synonym 3, 4-Dichloro-N-(1-methylbutyl)benzamide
Chemical Name or Material NSC 405020
Assay Percent Range >99%
Molecular Formula C12H15Cl2NO
CAS 7497-07-6
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature