Please login to your online account to display your discounted pricing

Tocris™ NSC 3852

Histone deacetylase inhibitor

$75.35 - $257.85

Specifications

Product Type NSC 3852
Inhibitors NSC 3852
Molecular Weight 174.16
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

252110

 
tocris™
RDS252110
10mg Each for $75.35

252150

 
tocris™
RDS252150
50mg Each for $257.85
Description & Specifications

Specifications

Product Type NSC 3852
Inhibitors NSC 3852
Molecular Weight 174.16
Purity >98

Histone deacetylase inhibitor. Causes cell differentiation and antiproliferative activity in MCF-7 human breast cancer cells in vitro and displays antitumor activity in vivo.

  • Chemical Name: 5-Nitroso-8-quinolinol
  • Formula: C9H6N2O2
  • CAS Number: 3565-26-2
  • Solubility: Soluble to 100mM in DMSO and to 10mM in ethanol