Please login to your online account to display your discounted pricing

NSC 33994, Tocris™

JAK2 inhibitor

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material NSC 33994
Assay Percent Range >98%
Molecular Formula C28H42N2O2
Formula Weight 438.65
Synonym E-4,4'-(1,2-Diethyl-1,2-ethenediyl)bis[2-[(diethylamino)methyl]phenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

433810

 
r & d systems
RDS433810
10mg Each for $244.81

433850

 
r & d systems
RDS433850
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 438.65
Quantity 10mg
Assay Percent Range >98%
CAS 82058-16-0
Chemical Name or Material NSC 33994
Molecular Formula C28H42N2O2
Synonym E-4,4'-(1,2-Diethyl-1,2-ethenediyl)bis[2-[(diethylamino)methyl]phenol
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C