Please login to your online account to display your discounted pricing

NSC 109555 ditosylate, Tocris

Selective Chk2 inhibitor

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material NSC 109555 ditosylate
Assay Percent Range >98%
Molecular Formula C19H24N10O·2C7H8O3S
Formula Weight 752.86
Synonym 4, 4'-diacetyldiphenylurea bis(guanylhydrazone) ditosylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

303450

 
r & d systems
RDS303450
50mg Each for $915.10

303410

 
r & d systems
RDS303410
10mg Each for $218.74
Description & Specifications

Specifications

Formula Weight 752.86
Quantity 50mg
Chemical Name or Material NSC 109555 ditosylate
Assay Percent Range >98%
Molecular Formula C19H24N10O·2C7H8O3S
CAS 66748-43-4
Synonym 4, 4'-diacetyldiphenylurea bis(guanylhydrazone) ditosylate
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C