Please login to your online account to display your discounted pricing

Alfa Aesar™ NS-398, 98%


Manufacturer: alfa aesar™  J67313MB

 View more versions of this product

Catalog No. AAJ67313MB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Assay 98%
Assay 98%
CAS 123653-11-2
Molecular Formula C13H18N2O5S
Formula Weight 314.36g/mol
Synonym N-[2-(Cyclohexyloxy)-4-nitrophenyl]-methanesulfonamide;
Quantity 25mg
Description Off-white
Format Solid