Please login to your online account to display your discounted pricing

NQDI 1, Tocris™

Inhibitor of apoptosis signal-regulating kinase 1 (ASK1)

$270.88 - $1,140.36

Chemical Identifiers

Chemical Name or Material NQDI 1
Assay Percent Range >99%
Molecular Formula C19H13NO4
Formula Weight 319.31
Synonym 2,7-Dihydro-2,7-dioxo-3H-naphtho[1,2,3-de]quinoline-1-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

442910

 
r & d systems
RDS442910
10mg Each for $270.88

442950

 
r & d systems
RDS442950
50mg Each for $1,140.36
Description & Specifications

Specifications

Formula Weight 319.31
Quantity 10mg
Chemical Name or Material NQDI 1
Assay Percent Range >99%
Molecular Formula C19H13NO4
CAS 175026-96-7
Synonym 2,7-Dihydro-2,7-dioxo-3H-naphtho[1,2,3-de]quinoline-1-carboxylic acid ethyl ester
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C