NPS Corp. Spilfyter™ Cellulose Sorbent Spill Kits
Quick spill control for large, non-aggressive spills
Manufacturer:  NPS Corp. JV152020
 View more versions of this product
Catalog No. 29-635-1097
Description
Description
  • Assortment of poly and overpack drums in different sizes
  • Cellulose-based sorbent offers different capacities of absorption
Specifications
Specifications
19 gal.
20 gal., 75.7L
Cellulose
75.7L
72L
Four 3 in. x 4 ft. cellulose socks, 24 8 x 18 in. cellulose pads, disposal bag and twist tie, 20 gal. poly drum with Lever-lock metal ring top
20 gal.
NPS Spill Kit
Documents