Liquid Safety Solutions MERCSORB™ Powder
Use for dangerous mercury spills
Manufacturer:  Liquid Safety Solutions JV522500
 View more versions of this product
Catalog No. 29-633-511
Includes: Mercsorb Amalgamation Powder (500g)
Description
Description
  • Amalgamation powder converts elemental mercury into amalgam
  • Stops dangerous mercury vapors from being emitted
Specifications
Specifications
5.5 lb., 2.5kg
2.5kg
2.5kg
Amalgam Powder
5.5 lb.
5.5 lb.
Converts elemental mercury into amalgam, stops dangerous mercury vapors from being emitted
SDS
Documents
To be used by trained personnel only