Liquid Safety Solutions Gapseal™ Plugging Compound
Temporary fix for leaking containers
Manufacturer:  Liquid Safety Solutions 780281
 View more versions of this product
Catalog No. 19-060-552
Description
Description
  • Ready-to-use compound temporarily stops nonaggressive and hydrocarbon-based leaks
  • Use to divert or contain spill and leaks from punctured drums, storage containers, and pipes until they can be properly drained or moved into a containment area
  • Gapseal Universal handles spills of aggressive, nonaggressive, water-, and hydrocarbon-based liquids
  • Gapseal Hydrocarbon is designed for use with hydrocarbon-based liquids only
  • Pre-mixed in 1 lb. containers
Gapseal is intended for use as a temporary method for stopping leaks and spills, and must be used in conjunction with secondary containment. Do not use Gapseal hydrocarbon for more than 12 hours or leave unattended. Not recommended for use with strong oxidizers, bases, or acids.
Specifications
Specifications
Used for plugging punctured drums or vessels
1 Case
907.18g
Gapseal™ Plugging Compound
2 lb.