Please login to your online account to display your discounted pricing

NPS ALX Compound 4a dihydrochloride, Tocris

Potent and selective 5-HT6 antagonist

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material NPS ALX Compound 4a dihydrochloride
Assay Percent Range >99%
Molecular Formula C25H25N3O2S·2HCl
Formula Weight 504.47
Synonym 6-(Hexahydropyrrolo[1, 2-a]pyrazin-2(1H)-yl)-1-(1-naphthalenylsulfonyl)-1H-indole dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

328510

 
r & d systems
RDS328510
10mg Each for $244.81

328550

 
r & d systems
RDS328550
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 504.47
Quantity 10mg
Chemical Name or Material NPS ALX Compound 4a dihydrochloride
Molecular Formula C25H25N3O2S·2HCl
Synonym 6-(Hexahydropyrrolo[1, 2-a]pyrazin-2(1H)-yl)-1-(1-naphthalenylsulfonyl)-1H-indole dihydrochloride
Recommended Storage Store at +4°C
CAS 299433-10-6
Assay Percent Range >99%
Safety and Handling

Store at +4°C