Please login to your online account to display your discounted pricing

NPEC-caged-serotonin, Tocris™

Caged serotonin

$238.81 - $984.19

Chemical Identifiers

Chemical Name or Material NPEC-caged-serotonin
Assay Percent Range >99%
Molecular Formula C19H19N3O5
Formula Weight 369.37
Synonym 3-((N)-1-(2-Nitrophenyl)ethylcarboxy)-(2-Aminoethyl)-1H-indol-5-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

399150

 
r & d systems
399150
50mg Each for $984.19

399110

 
r & d systems
399110
10mg Each for $238.81
Description & Specifications

Specifications

Formula Weight 369.37
Quantity 50mg
Chemical Name or Material NPEC-caged-serotonin
Molecular Formula C19H19N3O5
Assay Percent Range >99%
CAS 1257326-22-9
Synonym 3-((N)-1-(2-Nitrophenyl)ethylcarboxy)-(2-Aminoethyl)-1H-indol-5-ol
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C