Please login to your online account to display your discounted pricing

NPEC-caged-(S)-AMPA, Tocris™

Caged (S)-AMPA

$1,563.12 - $1,563.12

Chemical Identifiers

Chemical Name or Material NPEC-caged-(S)-AMPA
Assay Percent Range >98%
Molecular Formula C16H17N3O8
Formula Weight 379.32
Synonym (N)-1-(2-Nitrophenyl)ethylcarboxy-(S)-α-1-(2-nitrophenyl)ethylcarboxyamino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

333050

 
r & d systems
RDS333050
50mg Each for $1,563.12
Description & Specifications

Specifications

Formula Weight 379.32
Quantity 50mg
Chemical Name or Material NPEC-caged-(S)-AMPA
Molecular Formula C16H17N3O8
CAS 1257323-84-4
Synonym (N)-1-(2-Nitrophenyl)ethylcarboxy-(S)-α-1-(2-nitrophenyl)ethylcarboxyamino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
Recommended Storage Store at -20°C
Assay Percent Range >98%
Safety and Handling

Store at -20°C