Please login to your online account to display your discounted pricing

NPEC-caged-noradrenalin, Tocris™

Caged noradrenalin

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material NPEC-caged-noradrenalin
Assay Percent Range >98%
Molecular Formula C17H18N2O7
Formula Weight 362.33
Synonym 4-[(1R)-[2-[1-(2-Nitrophenyl)ethyl]carboxyamino]-1-hydroxyethyl]benzene-1, 2-diol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

399010

 
r & d systems
RDS399010
10mg Each for $257.85

399050

 
r & d systems
RDS399050
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 362.33
Quantity 10mg
Chemical Name or Material NPEC-caged-noradrenalin
Assay Percent Range >98%
Molecular Formula C17H18N2O7
CAS 1257323-83-3
Synonym 4-[(1R)-[2-[1-(2-Nitrophenyl)ethyl]carboxyamino]-1-hydroxyethyl]benzene-1, 2-diol
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C