Please login to your online account to display your discounted pricing

NPEC-caged-D-AP5, Tocris™

Caged D-AP5

$316.20 - $1,307.92

Chemical Identifiers

Chemical Name or Material NPEC-caged-D-AP5
Assay Percent Range >99%
Molecular Formula C14H19N2O9P
Formula Weight 390.28
Synonym D-[[1-(2-Nitrophenyl)ethyl]carbamoyl]-2-amino]-5-phosphonopentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

423010

 
r & d systems
RDS423010
10mg Each for $316.20

423050

 
r & d systems
RDS423050
50mg Each for $1,307.92
Description & Specifications

Specifications

Formula Weight 390.28
Quantity 10mg
Synonym D-[[1-(2-Nitrophenyl)ethyl]carbamoyl]-2-amino]-5-phosphonopentanoic acid
Recommended Storage Store at -20°C
Molecular Formula C14H19N2O9P
CAS 1416943-27-5
Assay Percent Range >99%
Chemical Name or Material NPEC-caged-D-AP5
Safety and Handling

Store at -20°C