Please login to your online account to display your discounted pricing

NPEC-caged-(1S,3R)-ACPD, Tocris

Caged (1S,3R)-ACPD

$388.20 - $1,596.86

Chemical Identifiers

Chemical Name or Material NPEC-caged-(1S, 3R)-ACPD
Assay Percent Range >98%
Molecular Formula C16H18N2O8
Formula Weight 366.32
Synonym (1S, 3R)-1-(2-Nitrophenyl)ethylcarboxyaminocyclopentane-1, 3-dicarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

333110

 
r & d systems
RDS333110
10mg Each for $388.20

333150

 
r & d systems
RDS333150
50mg Each for $1,596.86
Description & Specifications

Specifications

Formula Weight 366.32
Quantity 10mg
Chemical Name or Material NPEC-caged-(1S, 3R)-ACPD
CAS 1315379-60-2
Recommended Storage Desiccate at Room Temperature
Assay Percent Range >98%
Synonym (1S, 3R)-1-(2-Nitrophenyl)ethylcarboxyaminocyclopentane-1, 3-dicarboxylic acid
Molecular Formula C16H18N2O8
Safety and Handling

Desiccate at Room Temperature