missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ NP-40 lysis buffer, high salt

Specifications

Chemical Name or Material NP-40 lysis buffer
Name Note high salt
Formulation 50mM Tris-HCl (pH 7.4), 500mM NaCl, 1% NP-40 and 5mM EDTA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61428AK View Documents Alfa Aesar™
J61428AK
250mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ61428AP View Documents Alfa Aesar™
J61428AP
500mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

NP-40 lysis buffer
high salt
50mM Tris-HCl (pH 7.4), 500mM NaCl, 1% NP-40 and 5mM EDTA