missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ vH RAS Protein

$296.67 - $692.88

Specifications

Conjugate Unlabeled
For Use With (Application) Immunoassay, SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP15083101 Novus Biologicals Canada
NBP15083101MG
0.1mg Each for $296.67
NBP1508315 Novus Biologicals Canada
NBP15083105MG
0.5mg Each for $692.88
Specifications Description & Specifications

Specifications

Unlabeled
Immunoassay, SDS-Page
Store at -80°C. Avoid freeze-thaw cycles.
RUO