missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ VEGF Protein

$1,044.00 - $2,309.00

Specifications

Conjugate Unlabeled
For Use With (Application) Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP1428915 Novus Biologicals Canada
NBP14289105MG
0.5mg Each for $2,309.00
NBP14289101 Novus Biologicals Canada
NBP14289101MG
0.1mg Each for $1,044.00
Specifications Description & Specifications

Specifications

Unlabeled
Immunoassay
Store at -80°C. Avoid freeze-thaw cycles.
RUO