missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ Troponin T Type 2 (cardiac) Protein

$385.90 - $1,111.00

Specifications

Conjugate Unlabeled
For Use With (Application) ELISA, SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBC1287655 Novus Biologicals Canada
NBC12876505MG
0.5mg Each for $1,111.00
NBC12876501 Novus Biologicals Canada
NBC12876501MG
0.1mg Each for $385.90
Specifications Description & Specifications

Specifications

Unlabeled
ELISA, SDS-Page
Store at -80°C. Avoid freeze-thaw cycles.
RUO