missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ Transaldolase 1 Protein

$329.00 - $802.44

Specifications

Conjugate Unlabeled
For Use With (Application) SDS-Page
Storage Requirements Store at 4°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP1453165 Novus Biologicals Canada
NBP14531605MG
0.5mg Each for $802.44
NBP14531601 Novus Biologicals Canada
NBP14531601MG
0.1mg Each for $329.00
Specifications Description & Specifications

Specifications

Unlabeled
SDS-Page
Store at 4°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
RUO