Please login to your online account to display your discounted pricing

Novus Biologicals™ TRAIL/TNFSF10 Protein


Manufacturer: novus biologicals canada  NBC118505005MG

 View more versions of this product

Catalog No. NBC11850505

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 23kDa
Purification Method Protein
Purity >95%
Quantity 0.05mg
Source Human
Immunogen MVRERGPQRVAAHITGTRGRSNTLSSPNSKNEKALGRKINSWESSRSGHSFLSNLHLRNGELVIHEKGFYYIYSQTYFRFQEEIKENTKNDKQMVQYIYKYTSYPDPILLMKSARNSCWSKDAEYGLYSIYQGGIFELKENDRIFVSVTNEHLIDMDHEASFFGAFLVG
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Concentration 1.0mg/mL
Common Name TRAIL/TNFSF10
Conjugate Unlabeled
For Use With (Application) ELISA, SDS-Page
Formulation Liquid in 20mM Tris pH 7.5, 300mM NaCl, 0.1mM DTT, 10% glycerol