missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ TNF-alpha Protein

$279.41 - $1,924.00

Specifications

Conjugate Unlabeled
For Use With (Application) ELISA, Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14289901 Novus Biologicals Canada
NBP142899001MG
0.01mg Each for $279.41
NBP1428991M Novus Biologicals Canada
NBP14289901MG
0.1mg Each for $659.00
NBP1428995 Novus Biologicals Canada
NBP14289905MG
0.5mg Each for $1,924.00
Specifications Description & Specifications

Specifications

Unlabeled
ELISA, Immunoassay
Store at -80°C. Avoid freeze-thaw cycles.
RUO