Please login to your online account to display your discounted pricing

Novus Biologicals™ SNX1 Protein

$299.66 - $802.44

Specifications

Conjugate Unlabeled
For Use With (Application) ELISA, SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14834705

 
novus biologicals canada
NBP148347005MG
0.05mg Each for $802.44

NBP14834701

 
novus biologicals canada
NBP148347001MG
0.01mg Each for $299.66
Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) ELISA, SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO