missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ PRMT1 Protein

$254.34 - $254.34

Specifications

Conjugate Unlabeled
For Use With (Application) ELISA, Functional Assay, SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBC11844501 Novus Biologicals Canada
NBC11844501MG
0.1mg Each for $254.34
NBC11844601 Novus Biologicals Canada
NBC11844601MG
0.1mg Each for $254.34
Specifications Description & Specifications

Specifications

Unlabeled
ELISA, Functional Assay, SDS-Page
Store at -80°C. Avoid freeze-thaw cycles.
RUO