Please login to your online account to display your discounted pricing

Novus Biologicals Polyinosinic-polycytidylic Acid


For activation of TLR3

Manufacturer: novus biologicals  NBP22528850MG

 View more versions of this product

Catalog No. NBP22528850

  • / Each
 

Description & Specifications

Specifications

Product Type Polyinosinic-polycytidylic Acid
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 50mg
For Use With (Application) For activation of TLR3

Polyinosinic-polycytidylic acid (Poly(I:C)) is a synthetic analog of double stranded RNA (dsRNA) that is produced during the replicative life cycle of a majority of viruses. Poly(I:C) is known to induce type-1 interferon (IFN) through TLR3. Poly(I:C) directly binds to TLR3.

Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.