Please login to your online account to display your discounted pricing

Novus Biologicals™ PIN4 Protein

$385.90 - $1,066.00

Specifications

Conjugate Unlabeled
For Use With (Application) SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1510685

 
novus biologicals canada
NBP15106805MG
0.5mg Each for $1,066.00

NBP15106801

 
novus biologicals canada
NBP15106801MG
0.1mg Each for $385.90
Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO