missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ PHPT1 Protein

$299.66 - $802.44

Specifications

Conjugate Unlabeled
For Use With (Application) ELISA, SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP1494395 Novus Biologicals Canada
NBP14943905MG
0.5mg Each for $802.44
NBP14943901 Novus Biologicals Canada
NBP14943901MG
0.1mg Each for $299.66
Specifications Description & Specifications

Specifications

Unlabeled
ELISA, SDS-Page
Store at -80°C. Avoid freeze-thaw cycles.
RUO