missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ PDGF-B Protein

$222.95 - $280.88

Specifications

Conjugate Unlabeled
For Use With (Application) Functional Assay, Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14318003 Novus Biologicals Canada
NBP1431800025MG
0.025mg Each for $280.88
NBP14318005 Novus Biologicals Canada
NBP1431800005MG
0.005mg Each for $222.95
Specifications Description & Specifications

Specifications

Unlabeled
Functional Assay, Immunoassay
Store at -80°C. Avoid freeze-thaw cycles.
RUO