missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ OGG1 Protein

$329.00 - $802.44

Specifications

Conjugate Unlabeled
For Use With (Application) ELISA, In vitro assay, SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14531801 Novus Biologicals Canada
NBP14531801MG
0.1mg Each for $329.00
NBP1453185 Novus Biologicals Canada
NBP14531805MG
0.5mg Each for $802.44
Specifications Description & Specifications

Specifications

Unlabeled
ELISA, In vitro assay, SDS-Page
Store at -80°C. Avoid freeze-thaw cycles.
RUO