Please login to your online account to display your discounted pricing

Novus Biologicals™ NUDT9 Protein

$299.66 - $802.44

Specifications

Conjugate Unlabeled
For Use With (Application) SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17230901

 
novus biologicals canada
NBP17230901MG
0.1mg Each for $299.66

NBP1723095

 
novus biologicals canada
NBP17230905MG
0.5mg Each for $802.44
Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO