missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ MDH2 Protein

$299.66 - $802.44

Specifications

Conjugate Unlabeled
For Use With (Application) Western Blot, ELISA
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14832503 Novus Biologicals Canada
NBP1483250250MG
0.25mg Each for $802.44
NBP14832505 Novus Biologicals Canada
NBP148325005MG
0.05mg Each for $299.66
Specifications Description & Specifications

Specifications

Unlabeled
Western Blot, ELISA
Store at -80°C. Avoid freeze-thaw cycles.
RUO