missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ M-CSF Protein

$164.00 - $279.41

Specifications

Conjugate Unlabeled
For Use With (Application) Flow Cytometry, Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14316603 Novus Biologicals Canada
NBP1431660025MG
0.025mg Each for $279.41
NBP14316605 Novus Biologicals Canada
NBP1431660005MG
0.005mg Each for $164.00
Specifications Description & Specifications

Specifications

Unlabeled
Flow Cytometry, Immunoassay
Store at -80°C. Avoid freeze-thaw cycles.
RUO