Please login to your online account to display your discounted pricing
Novus Biologicals™ Lightning-Link™ Type B Biotin Antibody Labeling Kit
Antibody labeling kit
$300.00 - $659.00
Specifications
Product Type Type B Biotin Antibody Labeling Kit
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
7150010
novus biologicals™
7150010
3 x 200μg Reactions Each for $527.00
7150030
novus biologicals™
7150030
3 x 20μg Reactions Each for $300.00
7150015
novus biologicals™
7150015
1 x 2mg Reaction Each for $659.00
Description & Specifications
Specifications
Product Type Type B Biotin Antibody Labeling Kit
The attachment of biotin to biomolecules is an important laboratory technique. Biotin binds to the tetrameric avidin proteins, including streptavidin and neutravidin, with exceptionally high affinity, and this interaction is exploited in various applications such as western blotting, immunohisthochemistry and ELISA. Our biotinylation kits are optimised to create high quality conjugates and minimise assay background. There is also no need for a wash or desalting step following the biotinylation reaction, saving you both time and material. Lightning-Link Biotin has been optimised for two separate applications: Type A - intended for assays in which a streptavidin-labeled detection reagent will be used. Type B - optimised for assays in which the biotinylated protein is captured by streptavidin immobilized on a surface (ie plates, nitrocellulose, magnetic beads etc).