Please login to your online account to display your discounted pricing
Novus Biologicals™ Lightning-Link™ Rapid FluoProbe™594 Antibody Labeling Kit
Antibody labeling kit
$386.00 - $837.00
Specifications
Product Type Rapid FluoProbe594 Antibody Labeling Kit
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
3610010
novus biologicals canada
361-0010
3 x 200μg Reactions Each for $483.00
3610015
novus biologicals canada
361-0015
1 x 2mg Reaction Each for $837.00
3610030
novus biologicals canada
361-0030
3 x 20μg Reactions Each for $386.00
Description & Specifications
Specifications
Product Type Rapid FluoProbe594 Antibody Labeling Kit
FluoProbes 594 is one of a new generation of fluorescent labels designed for labelling biomolecules. It has a strong absorption at 591nm, high fluorescence at 616nm (extinction coefficient 9.2 x 104 cm-1M-1) and high quantum yield. FluoProbes 594 can be used as a substitute dye for BODIPY TR-x, Carboxynaphofluorescein and Alexa Fluor 594.