Please login to your online account to display your discounted pricing
Novus Biologicals™ Lightning-Link™ Rapid Atto594 Antibody Labeling Kit
Antibody labeling kit
$406.00 - $879.00
Specifications
Product Type Rapid Atto594 Antibody Labeling Kit
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
3520015
novus biologicals canada
3520015
1 x 2mg Reaction Each for $879.00
3520030
novus biologicals canada
3520030
3 x 20μg Reactions Each for $406.00
Description & Specifications
Specifications
Product Type Rapid Atto594 Antibody Labeling Kit
Atto594 is a fluorescent label belonging to the Rhodamine class of dyes. The dye has a strong absorption at 601nm, high fluorescence at 627nm (extinction coefficient 1.2 x105 cm-1M-1) and quantum yield (0.85).