Please login to your online account to display your discounted pricing
Novus Biologicals™ Lightning-Link™ Rapid Atto 700 Antibody Labeling Kit
Antibody labeling kit
$332.00 - $805.00
Specifications
Product Type Rapid Atto 700 Antibody Labeling Kit
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
3540010
novus biologicals canada
3540010
3 x 200μg Reactions Each for $527.00
3540015
novus biologicals canada
3540015
1 x 2mg Reaction Each for $805.00
3540030
novus biologicals canada
3540030
3 x 20μg Reactions Each for $332.00
Description & Specifications
Specifications
Product Type Rapid Atto 700 Antibody Labeling Kit
Atto700 is one of a new generation of fluorescent labels that emits in the red spectral region. It has a strong absorption peak at 690nm, with an extinction coefficient of 1.2 x 105 cm-1M-1, and a maximum emission peak at 725nm.