Please login to your online account to display your discounted pricing
Novus Biologicals™ Lightning-Link™ Avidin Antibody Labeling Kit
Antibody labeling kit
$300.00 - $493.00
Specifications
Product Type Avidin Antibody Labeling Kit
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
7170010
novus biologicals canada
717-0010
3 x 100μg Reactions Each for $493.00
7170030
novus biologicals canada
717-0030
3 x 0.01mg Reactions Each for $300.00
Description & Specifications
Specifications
Product Type Avidin Antibody Labeling Kit
Avidin is a 66kDa tetrameric glycoprotein composed of four identical subunits, which has widespread applications due to its very high affinity to the vitamin biotin.