Please login to your online account to display your discounted pricing

Novus Biologicals™ LC3B/MAP1LC3B Protein

$299.66 - $802.44

Specifications

Conjugate Unlabeled
For Use With (Application) SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP1509605

 
novus biologicals canada
NBP15096005MG
0.5mg Each for $802.44

NBP15096001

 
novus biologicals canada
NBP15096001MG
0.1mg Each for $299.66
Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO