missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ KRAS Protein

$296.59 - $692.88

Specifications

Conjugate Unlabeled
For Use With (Application) ELISA, Immunoassay, SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP1452745 Novus Biologicals Canada
NBP14527405MG
0.5mg Each for $692.88
NBP14527401 Novus Biologicals Canada
NBP14527401MG
0.1mg Each for $296.59
Specifications Description & Specifications

Specifications

Unlabeled
ELISA, Immunoassay, SDS-Page
Store at -80°C. Avoid freeze-thaw cycles.
RUO