missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ IL-9 Protein

$276.20 - $516.00

Specifications

Conjugate Unlabeled
For Use With (Application) ELISA, Flow Cytometry, Immunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14316203 Novus Biologicals Canada
NBP1431620025MG
0.025mg Each for $516.00
NBP14316205 Novus Biologicals Canada
NBP1431620005MG
0.005mg Each for $276.20
Specifications Description & Specifications

Specifications

Unlabeled
ELISA, Flow Cytometry, Immunoassay
Store at -80°C. Avoid freeze-thaw cycles.
RUO